Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioflavin T, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2390-54-7
C17H19ClN2S
318.863
MFCD00011944
JADVWWSKYZXRGX-UHFFFAOYSA-M
Basic Yellow 1,C.I. 49005
16953
CHEBI:76023
4-(3,6-dimethyl-1,3-benzothiazol-3-ium-2-yl)-N,N-dimethylaniline;chloride
CC1=CC2=C(C=C1)[N+](=C(S2)C3=CC=C(C=C3)N(C)C)C.[Cl-]
2390-54-7
C17H19ClN2S
318.863
MFCD00011944
JADVWWSKYZXRGX-UHFFFAOYSA-M
Basic Yellow 1,C.I. 49005
16953
CHEBI:76023
4-(3,6-dimethyl-1,3-benzothiazol-3-ium-2-yl)-N,N-dimethylaniline;chloride
CC1=CC2=C(C=C1)[N+](=C(S2)C3=CC=C(C=C3)N(C)C)C.[Cl-]
Beilstein 27, 377
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most organic solvent
Infrared Spectrum Authentic
Formula Weight 318.85
Physical Form Powder
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in most organic solvent
UV Spectrometry Complies
Color Ochre-Yellow to Yellow
Quantity 25g
Chemical Name or Material Thioflavin T
Show More Show Less

EINECSNumber : 219-228-9

TSCA : TSCA