Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thioisobutyramide, 95%

Click to view available options
Quantity:
1g
5g
25g
13515-65-6
C4H9NS
103.183
MFCD07369538
NPCLRBQYESMUPD-UHFFFAOYSA-N
thioisobutyramide,isobutylthioamide,isobutyrthioamide,2-methylpropaneothioamide,2-methylpropane-thioamide,2-methyl-propanethioamide,acmc-209by0,propanethioamide,2-methyl,ksc519m8d,2-methyl propanethioamide
10909507
2-methylpropanethioamide
CC(C)C(=S)N
13515-65-6
C4H9NS
103.183
MFCD07369538
NPCLRBQYESMUPD-UHFFFAOYSA-N
thioisobutyramide,isobutylthioamide,isobutyrthioamide,2-methylpropaneothioamide,2-methylpropane-thioamide,2-methyl-propanethioamide,acmc-209by0,propanethioamide,2-methyl,ksc519m8d,2-methyl propanethioamide
10909507
2-methylpropanethioamide
CC(C)C(=S)N
Quantity 25g
Melting Point 45°C to 48°C
Linear Formula (CH3)2CHC(=S)NH2
Beilstein 1098427
Formula Weight 103.18
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Thioisobutyramide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C