Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thioisonicotinamide, 98%

Click to view available options
Quantity:
1g
5g
25g
2196-13-6
C6H6N2S
138.188
MFCD00006437
KPIIGXWUNXGGCP-UHFFFAOYSA-N
thioisonicotinamide,4-pyridinecarbothioamide,isothionicotinamide,4-pyridylthiocarboxamide,4-pyridylthioamide,thio-4-pyridinecarboxamide,isonicotinic acid thioamide,pyridine-4-thioamide,4-pyridinecarboxamide, thio,isonicotinamide, thio
2723788
pyridine-4-carbothioamide
C1=CN=CC=C1C(=S)N
2196-13-6
C6H6N2S
138.188
MFCD00006437
KPIIGXWUNXGGCP-UHFFFAOYSA-N
thioisonicotinamide,4-pyridinecarbothioamide,isothionicotinamide,4-pyridylthiocarboxamide,4-pyridylthioamide,thio-4-pyridinecarboxamide,isonicotinic acid thioamide,pyridine-4-thioamide,4-pyridinecarboxamide, thio,isonicotinamide, thio
2723788
pyridine-4-carbothioamide
C1=CN=CC=C1C(=S)N
Melting Point 198°C to 202°C (decomposition)
Quantity 1g
Beilstein 2174
Formula Weight 138.19
Percent Purity 98%
Chemical Name or Material Thioisonicotinamide

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : US4204000

TSCA : TSCA