Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioacetic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
507-09-5
C2H4OS
76.11
MFCD00004853
DUYAAUVXQSMXQP-UHFFFAOYSA-N
thioacetic acid,ethanethioic acid,thiacetic acid,thioacetic s-acid,acetyl mercaptan,ethanethiolic acid,thiolacetic acid,thionoacetic acid,methanecarbothiolic acid,ch3cosh
10484
CHEBI:16555
ethanethioic S-acid
CC(=O)S
507-09-5
C2H4OS
76.11
MFCD00004853
DUYAAUVXQSMXQP-UHFFFAOYSA-N
thioacetic acid,ethanethioic acid,thiacetic acid,thioacetic s-acid,acetyl mercaptan,ethanethiolic acid,thiolacetic acid,thionoacetic acid,methanecarbothiolic acid,ch3cosh
10484
CHEBI:16555
ethanethioic S-acid
CC(=O)S
Specific Gravity 1.07
Density 1.0700g/mL
Infrared Spectrum Authentic
Quantity 2.5kg
Melting Point -17.0°C
Color Colorless to Yellow
Refractive Index 1.4630 to 1.4650
Boiling Point 87.0°C to 93.0°C
Packaging Glass bottle
Flash Point 18°C
Assay Percent Range 98%
Linear Formula CH3COSH
Beilstein 02, 230
Fieser 01,1154
Merck Index 15, 9473
Solubility Information Solubility in water: 27g/L water (15°C) (hydrolyse). Other solubilities: soluble in alcohol and ether
Formula Weight 76.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Thioacetic acid, 98%
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 208-063-8

RTECSNumber : AJ5600000

TSCA : TSCA