Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ ThiolTracker™ Violet (glutathione detection reagent) Non-distribution product as customer accommodation.

ThiolTracker™ Violet (glutathione detection reagent)

$267.00 - $405.00

Specifications

Color Violet
Detection Method Fluorescent
Excitation Wavelength Range 405nm
For Use With (Application) For cell viability, proliferation and function, cellular imaging, flow cytometry
For Use With (Equipment) For flow cytometer, fluorescence microscope
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
T10095 Non-distribution product as customer accommodation. View Documents Molecular Probes™
T10095
180 rxn(s) Each for $405.00
Add to cart
 
T10096 Non-distribution product as customer accommodation. View Documents Molecular Probes™
T10096
5 Microplate(s) Each for $267.00
Add to cart
 
Description

Description

ThiolTracker™ Violet is a brighter and more robust intracellular thiol probe for reproducible GSH detection. Since reduced glutathione represents the majority of intracellular free thiols in the cell, ThiolTracker™ Violet can be used in estimating the cellular level of reduced glutathione.
ThiolTracker™ Violet offers the following;
  • 10X brighter than mBCL
  • UV or 405 nm excitation, 525 nm emission
  • Easy to use
  • Reproducible results
  • Can be fixed with aldehydes and permeabilized by Triton™ X-100 (0.5%)
  • Formaldehyde fixable and suvives detergent extraction
  • Can be used in multiplex assays including cytotoxicity studies
    Subcellular detection and localization of GSH is important in understanding the modulation of redox status, the effect of drugs, and the mechanisms of detoxification. Furthermore, differences in GSH levels in response to oxidative stress in subpopulations of cells have been reported, underscoring the importance of detection methods amenable to flow cytometry and automated fluorescence detection. ThiolTracker™ Violet can be used for Flow Cytometry, HCS imaging, and epifluorescent microscopy.
  • Specifications

    Specifications

    Violet
    405nm
    For flow cytometer, fluorescence microscope
    ≤–20°C
    Fluorescent
    For cell viability, proliferation and function, cellular imaging, flow cytometry
    Slide(s), Tube(s)
    ThiolTracker™ Violet (Glutathione Detection Reagent)

    For Research Use Only. Not for use in diagnostic procedures.

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit