Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiomorpholine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
123-90-0
C4H9NS
103.19
MFCD00005974
BRNULMACUQOKMR-UHFFFAOYSA-N
thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
67164
CHEBI:36392
thiomorpholine
C1CSCCN1
123-90-0
C4H9NS
103.19
MFCD00005974
BRNULMACUQOKMR-UHFFFAOYSA-N
thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
67164
CHEBI:36392
thiomorpholine
C1CSCCN1
Refractive Index 1.5370 to 1.539
Flash Point 63°C
Boiling Point 175°C to 178°C
Specific Gravity 1.088
Density 1.0880g/mL
Packaging Glass bottle
Quantity 5g
Assay Percent Range 96% min. (GC)
Beilstein 27, 9
Merck Index 15, 9452
Solubility Information Solubility in water: miscible. Other solubilities: miscible with many organic liquids
Formula Weight 103.19
Percent Purity 97%
Chemical Name or Material Thiomorpholine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-660-2