Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thionin (Certified Biological Stain), Fisher Chemical
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
Specifications
Bio. Stain Comm. Certification | Pass Test |
Dye Content | Report % |
Formula Weight | 263.74 |
Grade | Certified Biological Stain |
Packaging | Glass Bottle |
Color Index No. | 52000 |
Color | Black |
Quantity | 25 g |
Physical Form | Solid |
Chemical Name or Material | Thionin |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction