Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophene-2-ethylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
30433-91-1
C6H9NS
127.21
MFCD00051495
HVLUYXIJZLDNIS-UHFFFAOYSA-N
thiophene-2-ethylamine,2-thiopheneethylamine,2-thiophen-2-yl ethanamine,thiopheneethanamine,2-2-thienyl ethylamine,2-thien-2-ylethanamine,2-thiopheneethanamine,thiopheneethylamine,2-thiophen-2-yl ethan-1-amine,2-2-aminoethyl thiophene
116521
2-thiophen-2-ylethanamine
C1=CSC(=C1)CCN
30433-91-1
C6H9NS
127.21
MFCD00051495
HVLUYXIJZLDNIS-UHFFFAOYSA-N
thiophene-2-ethylamine,2-thiopheneethylamine,2-thiophen-2-yl ethanamine,thiopheneethanamine,2-2-thienyl ethylamine,2-thien-2-ylethanamine,2-thiopheneethanamine,thiopheneethylamine,2-thiophen-2-yl ethan-1-amine,2-2-aminoethyl thiophene
116521
2-thiophen-2-ylethanamine
C1=CSC(=C1)CCN
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5490 to 1.5510
Quantity 1g
Boiling Point 103.0°C to 105.0°C (15.0 mmHg)
Color Colorless to Yellow
Assay Percent Range 97.5% min. (GC)
Formula Weight 127.21
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Thiophene-2-ethylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 250-196-9