Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thiophene-2-sulfonyl chloride, 97%

Click to view available options
Quantity:
5g
25g
100g
16629-19-9
C4H3ClO2S2
182.636
MFCD00005426
VNNLHYZDXIBHKZ-UHFFFAOYSA-N
2-thiophenesulfonyl chloride,2-thienylsulfonyl chloride,thiophene-2-sulphonyl chloride,2-thiophenesulfonylchloride,2-thienylsulfonylchloride,2-thiophene sulfonyl chloride,chloro-2-thienylsulfone,pubchem7737,acmc-209dtw,thiophenesulfonyl chloride
85518
thiophene-2-sulfonyl chloride
C1=CSC(=C1)S(=O)(=O)Cl
16629-19-9
C4H3ClO2S2
182.636
MFCD00005426
VNNLHYZDXIBHKZ-UHFFFAOYSA-N
2-thiophenesulfonyl chloride,2-thienylsulfonyl chloride,thiophene-2-sulphonyl chloride,2-thiophenesulfonylchloride,2-thienylsulfonylchloride,2-thiophene sulfonyl chloride,chloro-2-thienylsulfone,pubchem7737,acmc-209dtw,thiophenesulfonyl chloride
85518
thiophene-2-sulfonyl chloride
C1=CSC(=C1)S(=O)(=O)Cl
Applications
2-Thiophenesulfonyl chloride has been used in the preparation of 2-((trans-2-phenyl cyclopropyl)sulfonyl)thiophene

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents, bases.
Refractive Index 1.573
Sensitivity Moisture sensitive
Flash Point >110°C (230°F)
Quantity 100g
Boiling Point 80°C to 81°C (1mmHg)
Melting Point 29°C to 31°C
UN Number UN3261
Beilstein 123536
Solubility Information Reacts with water.
Formula Weight 182.65
Percent Purity 97%
Chemical Name or Material Thiophene-2-sulfonyl chloride
Show More Show Less

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 240-677-1

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen