Alfa Aesar™ Thiophene-2-thiocarboxamide, 97%

Manufacturer: Alfa Aesar™ B2522506

Quantity 5g

 View more versions of this product

Catalog No. AAB2522506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20300-02-1
    Molecular Formula C5H5NS2
    Formula Weight 143.23
    MDL Number MFCD00052890
    UN Number UN2811
    Beilstein 110159
    Chemical Name or Material Thiophene-2-thiocarboxamide

    Specifications

    Formula Weight 143.23
    Molecular Formula C5H5NS2
    MDL Number MFCD00052890
    CAS 20300-02-1
    UN Number UN2811
    Beilstein 110159
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Melting Point 106° to 109°C
    Quantity 5g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material Thiophene-2-thiocarboxamide
    Assay 97%

    Description

    Applications
    Thiophene-2-thiocarboxamide is used as a pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit