Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thiophene-2-thiol, contains dimer, 97% as monomer, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.247 |
Boiling Point | 62°C to 65°C (12 mmHg) |
Flash Point | 66°C (151°F) |
Odor | Unpleasant |
Refractive Index | 1.617 |
Quantity | 1 g |
Beilstein | 104650 |
Sensitivity | Air and light sensitive |
Formula Weight | 116.2 |
Percent Purity | 97% |
Show More |
Safety and Handling
Hazard Category | H227-H302+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H227-H332-H315-H319-H335 Toxic if swallowed. Combustible liquid. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Hazard Class: 9; Packaging Group: III |
EINECSNumber | 231-881-1 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction