Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophene, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
110-02-1
C4H4S
84.14
MFCD00005413
YTPLMLYBLZKORZ-UHFFFAOYSA-N
thiofuran,thiole,thiophen,thiotetrole,thiacyclopentadiene,divinylene sulfide,thiaphene,thiofuram,thiofurfuran,furan, thio
8030
CHEBI:30856
thiophene
C1=CSC=C1
110-02-1
C4H4S
84.14
MFCD00005413
YTPLMLYBLZKORZ-UHFFFAOYSA-N
thiofuran,thiole,thiophen,thiotetrole,thiacyclopentadiene,divinylene sulfide,thiaphene,thiofuram,thiofurfuran,furan, thio
8030
CHEBI:30856
thiophene
C1=CSC=C1
Specific Gravity 1.065
Melting Point -38.0°C
Color Undesignated
Density 1.0650g/mL
Boiling Point 84.0°C
Refractive Index 1.5270 to 1.5290
Packaging Glass bottle
Infrared Spectrum Authentic
Water 0.1% max. (K.F.)
Flash Point −9°C
Quantity 500g
Assay Percent Range 99+%
Beilstein 17, 29
Merck Index 15, 9506
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic solvents
Formula Weight 84.14
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Thiophene, 99+%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-729-4

RTECSNumber : XM7350000

TSCA : TSCA