Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thiophenol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass Bottle
Metal drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -15.0°C |
Color | Colorless to Yellow |
Density | 1.0780g/mL |
Boiling Point | 167.0°C to 169.0°C |
Flash Point | 50°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass Bottle |
Molecular Formula | C6H6S |
Refractive Index | 1.5880 to 1.5900 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Flammable liquid and vapor. May cause respiratory irritation. Causes skin irritation. Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Suspected of damaging fertility or the unbo |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wash face,hands and any exposed skin thorou |
EINECSNumber | 203-635-3 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction