Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophenol, 99%, ACROS Organics™

Click to view available options
Quantity:
1L
5L
Packaging:
Glass Bottle
Metal drum
108-98-5
C6H6S
110.17
MFCD00004826
RMVRSNDYEFQCLF-UHFFFAOYSA-N
thiophenol,phenyl mercaptan,mercaptobenzene,phenylthiol,phenylmercaptan,thiofenol,phenol, thio,thiophenate,thiofenol czech,benzene, mercapto
7969
CHEBI:48498
benzenethiol
C1=CC=C(C=C1)S
108-98-5
C6H6S
110.17
MFCD00004826
RMVRSNDYEFQCLF-UHFFFAOYSA-N
thiophenol,phenyl mercaptan,mercaptobenzene,phenylthiol,phenylmercaptan,thiofenol,phenol, thio,thiophenate,thiofenol czech,benzene, mercapto
7969
CHEBI:48498
benzenethiol
C1=CC=C(C=C1)S
Boiling Point 167.0°C to 169.0°C
Specific Gravity 1.078
Melting Point -15.0°C
Refractive Index 1.5880 to 1.5900
Color Colorless to Yellow
Density 1.0780g/mL
Quantity 5L
Flash Point 50°C
Packaging Metal drum
Viscosity 0.981 mm2/s (30°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6H5SH
Beilstein 06, 294
Fieser 06,585; 07,367; 09,465; 10,399; 12,485; 13,53; 16,327
Merck Index 15, 9508
Solubility Information Solubility in water: insoluble. Other solubilities: very soluble in alcohol, miscible with ether, benzene and cs2
Formula Weight 110.17
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Thiophenol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger