Thiophenol, 99+%

Manufacturer: Alfa Aesar A1591630

Quantity 250g

 View more versions of this product

Catalog No. AAA1591630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-98-5
    Molecular Formula C6H6S
    Formula Weight 110.18
    MDL Number MFCD00004826
    UN Number UN2337
    Beilstein 506523
    Merck Index 14,9355
    Chemical Name or Material Thiophenol

    Specifications

    Formula Weight 110.18
    Quantity 250g
    TSCA TSCA
    MDL Number MFCD00004826
    UN Number UN2337
    Beilstein 506523
    Merck Index 14,9355
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H318-H226-H361-H315-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Suspected of damaging fertility or the unb
    Health Hazard 3 GHS P Statement
    P260-P280-P305+P351+P338-P309-P310-P302+P352
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for seve
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHENYL MERCAPTAN
    RTECS Number DC0525000
    Physical Form Liquid
    Density 1.075
    Assay Percent Range ≥99%
    CAS 108-98-5
    Storage Note 1 Air sensitive
    Molecular Formula C6H6S
    Chemical Name or Material Thiophenol
    Assay ≥99%

    Description

    Applications
    Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

    Solubility
    Miscible with most organic solvents and aqueous base. Immiscible with water.

    Notes
    Incompatible with strong acids, bases and oxidizing agents.

    GHS H Statement
    H300-H310-H330-H318-H226-H361-H315-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Suspected of damaging fertility or the unb

    GHS P Statement
    P260-P280-P305+P351+P338-P309-P310-P302+P352
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for seve

    Air sensitive

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit