Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophosgene, 85%, Acros Organics™

Click to view available options
Quantity:
25g,25g
100g,100g
500g,500g
2.5kg,2.5kg
Packaging:
Glass bottle,Glass bottle
463-71-8
CCl2S
114.98
MFCD00004918
ZWZVWGITAAIFPS-UHFFFAOYSA-N
thiophosgene,carbonothioic dichloride,carbon chlorosulfide,thiocarbonyl chloride,thiocarbonic dichloride,dichlorothiocarbonyl,thiofosgen,phosgene, thio,thiokarbonylchlorid,carbonyl chloride, thio
10040
CHEBI:29366
thiocarbonyl dichloride
C(=S)(Cl)Cl
463-71-8
CCl2S
114.98
MFCD00004918
ZWZVWGITAAIFPS-UHFFFAOYSA-N
thiophosgene,carbonothioic dichloride,carbon chlorosulfide,thiocarbonyl chloride,thiocarbonic dichloride,dichlorothiocarbonyl,thiofosgen,phosgene, thio,thiokarbonylchlorid,carbonyl chloride, thio
10040
CHEBI:29366
thiocarbonyl dichloride
C(=S)(Cl)Cl
C-X bond formation reagent
Density 1.5080g/mL,1.5080g/mL
Quantity 2.5kg,2.5kg
Color Orange-Red to Red,Orange-Red to Red
Boiling Point 73.0°C,73.0°C
Refractive Index 1.5442,1.5442
Flash Point 62°C,62°C
Packaging Glass bottle,Glass bottle
Specific Gravity 1.508,1.508
Assay Percent Range 85%
Linear Formula ClCSCl
Beilstein 03,134
Fieser 05,667; 09,466
Solubility Information Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol
Formula Weight 114.98
Physical Form Liquid
Percent Purity 85%
Solubility Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol
Chemical Name or Material Thiophosgene
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger