Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophosgene, 85%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2.5kg
Packaging:
Glass bottle
463-71-8
CCl2S
114.98
MFCD00004918
ZWZVWGITAAIFPS-UHFFFAOYSA-N
thiophosgene,carbonothioic dichloride,carbon chlorosulfide,thiocarbonyl chloride,thiocarbonic dichloride,dichlorothiocarbonyl,thiofosgen,phosgene, thio,thiokarbonylchlorid,carbonyl chloride, thio
10040
CHEBI:29366
thiocarbonyl dichloride
C(=S)(Cl)Cl
463-71-8
CCl2S
114.98
MFCD00004918
ZWZVWGITAAIFPS-UHFFFAOYSA-N
thiophosgene,carbonothioic dichloride,carbon chlorosulfide,thiocarbonyl chloride,thiocarbonic dichloride,dichlorothiocarbonyl,thiofosgen,phosgene, thio,thiokarbonylchlorid,carbonyl chloride, thio
10040
CHEBI:29366
thiocarbonyl dichloride
C(=S)(Cl)Cl
C-X bond formation reagent
Assay Percent Range 80% min. (GC)
Linear Formula ClCSCl
Beilstein 03,134
Fieser 05,667; 09,466
Solubility Information Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol
Formula Weight 114.98
Physical Form Liquid
Percent Purity 85%
Density 1.5080g/mL
Packaging Glass bottle
Refractive Index 1.5442
Solubility Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol
Specific Gravity 1.508
Boiling Point 73.0°C
Color Orange-Red to Red
Quantity 100g
Flash Point 62°C
Chemical Name or Material Thiophosgene
Show More Show Less

GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
If skin i

GHS Signal Word: Danger

EINECSNumber : 207-341-6