Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thiophosphateester Rabbit anti-Non-species specific, Clone: SD2020, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267738
Description
Thiophosphateester Monoclonal antibody specifically detects Thiophosphateester in Non-species specific samples. It is validated for Immunoprecipitation.Specifications
Thiophosphateester | |
SD2020 | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Non-species specific |
Immunoprecipitation | |
1 mg/mL | |
Immunoprecipitation 1:10 - 1:500 | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
Recombinant protein | |
Monoclonal | |
Protein A purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title