Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophosphoryl-PmmH-3 Dendrimer, Generation 0.5, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
159213-45-3
C21H15O6PS
426.379
MFCD03458720
RBHJCKHGVILXFB-UHFFFAOYSA-N
4-bis 4-formylphenoxy phosphinothioyloxybenzaldehyde,thiophosphoryl-pmmh-3 dendrime,tris 4-formylphenoxy phosphine sulfide,4-bis 4-formylphenoxy phosphino thioyloxybenzaldehyde
7168226
4-bis(4-formylphenoxy)phosphinothioyloxybenzaldehyde
C1=CC(=CC=C1C=O)OP(=S)(OC2=CC=C(C=C2)C=O)OC3=CC=C(C=C3)C=O
159213-45-3
C21H15O6PS
426.379
MFCD03458720
RBHJCKHGVILXFB-UHFFFAOYSA-N
4-bis 4-formylphenoxy phosphinothioyloxybenzaldehyde,thiophosphoryl-pmmh-3 dendrime,tris 4-formylphenoxy phosphine sulfide,4-bis 4-formylphenoxy phosphino thioyloxybenzaldehyde
7168226
4-bis(4-formylphenoxy)phosphinothioyloxybenzaldehyde
C1=CC(=CC=C1C=O)OP(=S)(OC2=CC=C(C=C2)C=O)OC3=CC=C(C=C3)C=O
NMR Complies
Color White
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Formula Weight 426.39
Physical Form Powder
Chemical Name or Material Thiophosphoryl-PMMH-3 Dendrimer, Generation 0.5, Generation 0.5