Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophosphoryl-PmmH-3 Dendrimer, Generation 1.0, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
173612-58-3
C24H24Cl6N6O3P4S4
909.33
MFCD03458721
GHSPLWJCBVPQQG-UHFFFAOYSA-N
thiophosphoryl-pmmh-3 dendrimer, generation 1.0
60029550
N-[[4-bis[4-[[dichlorophosphinothioyl(methyl)hydrazinylidene]methyl]phenoxy]phosphinothioyloxyphenyl]methylideneamino]-N-dichlorophosphinothioylmethanamine
CN(N=CC1=CC=C(C=C1)OP(=S)(OC2=CC=C(C=C2)C=NN(C)P(=S)(Cl)Cl)OC3=CC=C(C=C3)C=NN(C)P(=S)(Cl)Cl)P(=S)(Cl)Cl
173612-58-3
C24H24Cl6N6O3P4S4
909.33
MFCD03458721
GHSPLWJCBVPQQG-UHFFFAOYSA-N
thiophosphoryl-pmmh-3 dendrimer, generation 1.0
60029550
N-[[4-bis[4-[[dichlorophosphinothioyl(methyl)hydrazinylidene]methyl]phenoxy]phosphinothioyloxyphenyl]methylideneamino]-N-dichlorophosphinothioylmethanamine
CN(N=CC1=CC=C(C=C1)OP(=S)(OC2=CC=C(C=C2)C=NN(C)P(=S)(Cl)Cl)OC3=CC=C(C=C3)C=NN(C)P(=S)(Cl)Cl)P(=S)(Cl)Cl
Color White to Yellow
Quantity 500mg
Packaging Glass bottle
Infrared Spectrum Authentic
NMR Complies
Formula Weight 909.37
Physical Form Powder
Chemical Name or Material Thiophosphoryl-PMMH-3 Dendrimer, Generation 1.0, Generation 1.0