Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thioredoxin-1 Rabbit anti-Human, Clone: JF101-8, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267195
Description
Thioredoxin-1 Monoclonal antibody specifically detects Thioredoxin-1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
Thioredoxin-1 | |
JF101-8 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
TXN | |
Recombinant Protein | |
Protein A purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse, Rat |
Flow Cytometry, Western Blot | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry 1:50-1:100 | |
Purified | |
Rabbit | |
IgG | |
100μL | |
Primary | |
7295 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title