Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioredoxin Reductase 1/TRXR1 Mouse anti-Human, Clone: 30F1, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$337.00 - $482.50

Specifications

Antigen Thioredoxin Reductase 1/TRXR1
Clone 30F1
Concentration 1 mg/mL
Dilution Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25948950
SDP
View Documents
Novus Biologicals
NBP25948950UL
50 μL
Each for $337.00
Add to Cart
 
NBP259489X
SDP
View Documents
Novus Biologicals
NBP259489100UL
100 μL
Each for $482.50
Add to Cart
 
Description

Description

Thioredoxin Reductase 1/TRXR1 Monoclonal specifically detects Thioredoxin Reductase 1/TRXR1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Thioredoxin Reductase 1/TRXR1
1 mg/mL
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Breast Cancer, Lipid and Metabolism
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
7296
Recombinant human TXNRD1 (161-647aa) purified from E. coli
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
30F1
Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Monoclonal
Purified
RUO
Human
EC 1.8.1, EC 1.8.1.9, Gene associated with retinoic and IFN-induced mortality 12 protein, Gene associated with retinoic and interferon-induced mortality 12 protein, GRIM12, GRIM-12TR1, KDRF, KM-102-derived reductase-like factor, MGC9145, oxidoreductase, thioredoxin reductase 1, thioredoxin reductase 1, cytoplasmic, thioredoxin reductase GRIM-12, Thioredoxin reductase TR1, TR, Trxr1, TXNRTRXR1
TXNRD1
IgG2b κ
Protein A purified
SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit