Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioridazine Hydrochloride, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

T1426, 130-61-0, C21H26N2S2•HCl

Spectrum™ Thioridazine Hydrochloride, USP is an antipsychotic medicine. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Ignition Residue 0.001
Loss on Drying 0.004
Quantity 25 kg
Melting Range 159-165°C
pH 4.2 to 5.2
CAS 130-61-0
CAS Max % 1
Molecular Formula C21H27ClN2S2
InChI Key NZFNXWQNBYZDAQ-UHFFFAOYNA-N
SMILES [H+].[Cl-].CSC1=CC=C2SC3=CC=CC=C3N(CCC3CCCCN3C)C2=C1
IUPAC Name hydrogen 10-[2-(1-methylpiperidin-2-yl)ethyl]-2-(methylsulfanyl)-10H-phenothiazine chloride
Molecular Weight (g/mol) 407.03
Percent Purity 99 to 101%
Grade USP
Show More Show Less