Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiosalicylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
147-93-3
C7H6O2S
154.183
MFCD00004836
NBOMNTLFRHMDEZ-UHFFFAOYSA-N
thiosalicylic acid,2-mercaptobenzoic acid,o-mercaptobenzoic acid,2-thiosalicylic acid,o-thiosalicylic acid,2-carboxythiophenol,o-benzoic acid thiol,o-carboxythiophenol,o-sulfhydrylbenzoic acid,benzoic acid, 2-mercapto
5443
CHEBI:59124
2-sulfanylbenzoic acid
C1=CC=C(C(=C1)C(=O)O)S
147-93-3
C7H6O2S
154.183
MFCD00004836
NBOMNTLFRHMDEZ-UHFFFAOYSA-N
thiosalicylic acid,2-mercaptobenzoic acid,o-mercaptobenzoic acid,2-thiosalicylic acid,o-thiosalicylic acid,2-carboxythiophenol,o-benzoic acid thiol,o-carboxythiophenol,o-sulfhydrylbenzoic acid,benzoic acid, 2-mercapto
5443
CHEBI:59124
2-sulfanylbenzoic acid
C1=CC=C(C(=C1)C(=O)O)S
Assay Percent Range 98%
Linear Formula HSC6H4CO2H
Beilstein 10, 125
Merck Index 15, 9513
Formula Weight 154.18
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 164°C to 169°C
Color White to Yellow
Quantity 5g
Chemical Name or Material Thiosalicylic acid, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-704-3

RTECSNumber : DH3325000

TSCA : TSCA