Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiosemicarbazide, 98+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
79-19-6
CH5N3S
91.13
MFCD00007620
BRWIZMBXBAOCCF-UHFFFAOYSA-N
thiosemicarbazide,hydrazinecarbothioamide,n-aminothiourea,1-aminothiourea,semicarbazide, thio,isothiosemicarbazide,2-thiosemicarbazide,3-thiosemicarbazide,thiocarbamylhydrazine,thiocarbamoylhydrazine
2723789
aminothiourea
C(=S)(N)NN
79-19-6
CH5N3S
91.13
MFCD00007620
BRWIZMBXBAOCCF-UHFFFAOYSA-N
thiosemicarbazide,hydrazinecarbothioamide,n-aminothiourea,1-aminothiourea,semicarbazide, thio,isothiosemicarbazide,2-thiosemicarbazide,3-thiosemicarbazide,thiocarbamylhydrazine,thiocarbamoylhydrazine
2723789
aminothiourea
C(=S)(N)NN
Color White to Yellow
Infrared Spectrum Authentic
Melting Point 175.0°C to 180.0°C
Quantity 100g
Packaging Plastic bottle
Assay Percent Range 98+%
Linear Formula H2NCSNHNH2
Beilstein 03,195
Fieser 01,1164
Merck Index 15,9514
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 91.13
Physical Form Powder
Percent Purity 98+%
Chemical Name or Material Thiosemicarbazide, 99%
Show More Show Less

GHS H Statement
Fatal if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.

GHS Signal Word: Danger

EINECSNumber : 201-184-7

RTECSNumber : VT4200000

TSCA : TSCA