Alfa Aesar™ Thiotriazinone, 98+%

Manufacturer: Alfa Aesar™ B2266309

Quantity 10g

 View more versions of this product

Catalog No. AAB2266309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 58909-39-0
    Molecular Formula C4H5N3O2S
    Formula Weight 159.16
    MDL Number MFCD00185756
    Synonym Tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione
    Chemical Name or Material Thiotriazinone

    Specifications

    Formula Weight 159.16
    Synonym Tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione
    Quantity 10g
    Molecular Formula C4H5N3O2S
    MDL Number MFCD00185756
    CAS 58909-39-0
    Melting Point 168° to 171°C
    Chemical Name or Material Thiotriazinone
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit