Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Thiotriazinone, 98+%

Click to view available options
Quantity:
10g
50g
250g
58909-39-0
C4H5N3O2S
159.163
MFCD00185756
UMWWHOXOVPIGFD-UHFFFAOYSA-N
thiotriazinone,tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione,unii-679wf9q9w1,2-methyl-3-thioxo-1,2,4-triazinane-5,6-dione,1,2,4-triazin-2-methyl-6-hydroxy-3-thio-5-one,2-methyl-3-sulfanyl-1,2-dihydro-1,2,4-triazine-5,6-dione,1,2,4-triazine-5,6-dione, tetrahydro-2-methyl-3-thioxo,acmc-20ajjm,ceftriaxone impurity c,ksc269g8j
3034640
2-methyl-3-sulfanylidene-1,2,4-triazinane-5,6-dione
CN1C(=S)NC(=O)C(=O)N1
58909-39-0
C4H5N3O2S
159.163
MFCD00185756
UMWWHOXOVPIGFD-UHFFFAOYSA-N
thiotriazinone,tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione,unii-679wf9q9w1,2-methyl-3-thioxo-1,2,4-triazinane-5,6-dione,1,2,4-triazin-2-methyl-6-hydroxy-3-thio-5-one,2-methyl-3-sulfanyl-1,2-dihydro-1,2,4-triazine-5,6-dione,1,2,4-triazine-5,6-dione, tetrahydro-2-methyl-3-thioxo,acmc-20ajjm,ceftriaxone impurity c,ksc269g8j
3034640
2-methyl-3-sulfanylidene-1,2,4-triazinane-5,6-dione
CN1C(=S)NC(=O)C(=O)N1
Quantity 250g
Melting Point 168°C to 171°C
Formula Weight 159.16
Percent Purity ≥98%
Chemical Name or Material Thiotriazinone