Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiourea (Crystalline/Certified ACS), Fisher Chemical

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
62-56-6
CH4N2S
76.117
MFCD00008067
UMGDCJDMYOKAJW-UHFFFAOYSA-N
thiocarbamide,2-thiourea,pseudothiourea,isothiourea,sulourea,2-thiopseudourea,sulfourea,thiuronium,thiocarbonic acid diamide,urea, thio
2723790
CHEBI:36946
thiourea
C(=S)(N)N
62-56-6
CH4N2S
76.117
MFCD00008067
UMGDCJDMYOKAJW-UHFFFAOYSA-N
thiocarbamide,2-thiourea,pseudothiourea,isothiourea,sulourea,2-thiopseudourea,sulfourea,thiuronium,thiocarbonic acid diamide,urea, thio
2723790
CHEBI:36946
thiourea
C(=S)(N)N
Color White
Packaging Poly Bottle
Melting Point 176°C
Vapor Pressure 2.5mmHg at 25°C
Ignition Residue 0.1% max.
Quantity 2.5kg
Decomposition Information >180°C
Density 1.405g/cm³
Loss on Drying 0.5% max.
Identification Pass Test
Assay Percent Range ≥99 %
Linear Formula NH2CSNH2
UN Number 2811
Formula Weight 76.12
Physical Form Solid
Percent Purity ≥99%
Grade Certified ACS
Chemical Name or Material Thiourea
Show More Show Less

DOTInformation : DOT Class 6.1, : Poison