Alfa Aesar™ Thioxanthen-9-one, 98%

Manufacturer: Alfa Aesar™ B2376414

Quantity 25g

 View more versions of this product

Catalog No. AAB2376414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 492-22-8
    Molecular Formula C13H8OS
    Formula Weight 212.27
    MDL Number MFCD00005066
    Beilstein 140978
    Merck Index 14,9369
    Synonym Thioxanthone
    Chemical Name or Material Thioxanthen-9-one

    Specifications

    Formula Weight 212.27
    MDL Number MFCD00005066
    Merck Index 14,9369
    Melting Point 216° to 218°C
    Quantity 25g
    TSCA TSCA
    Molecular Formula C13H8OS
    CAS 492-22-8
    Beilstein 140978
    Synonym Thioxanthone
    Chemical Name or Material Thioxanthen-9-one
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit