Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THNSL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557258
Description
Immunogen sequence: AGYIAQKIGL PIRLVVAVNR NDIIHRTVQQ GDFSLSEAVK STLASAMDIQ VPYNMERVFW LLSGSDSQVT RALMEQFERT QSVNLPKEL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
THNSL2 (threonine synthase-like 2), also known as TSH2, is a 484 amino acid protein belonging to the threonine synthase family. Utilizing pyridoxal phosphate as a cofactor, THNSL2 may function as a catabolic phospholyase on gamma and beta phosphorylated substrates. THNSL2 may also degrade O-phospho-threonine to alpha-ketobutyrate, ammonia and phosphate. Existing as four alternatively spliced isoforms, THNSL2 is encoded by a gene mapping to human chromosome 2p11.2. As the second largest human chromosome, chromosome 2 makes up approximately 8% of the human genome and contains 237 million bases encoding over 1,400 genes. A number of genetic diseases are linked to genes on chromosome 2. Harlequin icthyosis, a rare skin deformity, is associated with mutations in the ABCA12 gene. The lipid metabolic disorder sitosterolemia is associated with ABCG5 and ABCG8 gene defects. An extremely rare recessive genetic disorder, Alstrom syndrome, is related to mutations in the ALMS1 gene.
Specifications
THNSL2 | |
Polyclonal | |
Unconjugated | |
THNSL2 | |
BC051244; FLJ10916; FLJ35504; RGD1309144; secreted osteoclastogenic factor of activated T cells; Secreted osteoclastogenic factor of activated T-cells; SOFAT; THNSL2; threonine synthase like 2; threonine synthase-like 2; threonine synthase-like 2 (bacterial); threonine synthase-like 2 (S. cerevisiae); THS2; TSH2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55258 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86YJ6 | |
THNSL2 | |
Recombinant protein corresponding to Human THNSL2. Recombinant protein control fragment (Product #RP-96115). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction