Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THOC3 Mouse anti-Human, Clone: OTI4H6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527131

Catalog No. PIMA527131


Add to cart

Description

Description

THOC3 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.TEX1 is part of the TREX (transcription/export) complex, which includes THO2 (MIM 300395), HPR1 (MIM 606930), ALY (MIM 604171), and UAP56 (MIM 606390).[supplied by OMIM].
Specifications

Specifications

THOC3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96J01
THO complex 3, TEX1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84321
Immunohistochemistry (Paraffin), Western Blot
OTI4H6
Unconjugated
THOC3
Liquid
THOC3
Human recombit protein fragment corresponding to amino acids 15-237 of THOC3 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit