Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THOC7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP2859100.1ML

Catalog No. NB033839


Add to cart

Description

Description

THOC7 Polyclonal specifically detects THOC7 in Human samples. It is validated for Western Blot.
Specifications

Specifications

THOC7
Unconjugated
Purified
Rabbit
Immunogen affinity purified
RUO
Primary
80145
Western Blot
PBS, 2% Sucrose
FLJ23445, fSAP24NIF3L1-binding protein 1, Functional spliceosome-associated protein 24, Ngg1 interacting factor 3 like 1 binding protein 1, Ngg1-interacting factor 3-like protein 1-binding protein 1, NIF3L1BP1hTREX30, THO complex 7 homolog (Drosophila), THO complex subunit 7 homolog
The immunogen is a synthetic peptide directed towards the C-terminal region of Human THOC7
0.1ml
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit