Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thorin I, pure, indicator grade, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass bottle
3688-92-4
C16H11AsN2Na2O10S2
576.289
MFCD00003888
ZQMPNLOZJUWHSA-WTBAQZMLSA-L
thoron,thoron ol,1-2-arsonophenylazo-2-naphthol-3,6-disulfonic acid disodium salt,apans,aspan,disodium 4e-4-2-arsonophenyl hydrazinylidene-3-oxonaphthalene-2,7-disulfonate,thoronol,o-3,6-disulfo-2-hydroxy-1-naphthyl-azo-benzenearsonic acid,2-2-hydroxy-3,6-disulfo-1-naphthylazo phenylarsonic acid disodium salt,2-3,6-disulfo-2-hydroxy-1-naphthylazo benzenearsonic acid disodium salt
6537679
disodium;(4E)-4-[(2-arsonophenyl)hydrazinylidene]-3-oxonaphthalene-2,7-disulfonate
C1=CC=C(C(=C1)NN=C2C3=C(C=C(C=C3)S(=O)(=O)[O-])C=C(C2=O)S(=O)(=O)[O-])[As](=O)(O)O.[Na+].[Na+]
3688-92-4
C16H11AsN2Na2O10S2
576.289
MFCD00003888
ZQMPNLOZJUWHSA-WTBAQZMLSA-L
thoron,thoron ol,1-2-arsonophenylazo-2-naphthol-3,6-disulfonic acid disodium salt,apans,aspan,disodium 4e-4-2-arsonophenyl hydrazinylidene-3-oxonaphthalene-2,7-disulfonate,thoronol,o-3,6-disulfo-2-hydroxy-1-naphthyl-azo-benzenearsonic acid,2-2-hydroxy-3,6-disulfo-1-naphthylazo phenylarsonic acid disodium salt,2-3,6-disulfo-2-hydroxy-1-naphthylazo benzenearsonic acid disodium salt
6537679
disodium;(4E)-4-[(2-arsonophenyl)hydrazinylidene]-3-oxonaphthalene-2,7-disulfonate
C1=CC=C(C(=C1)NN=C2C3=C(C=C(C=C3)S(=O)(=O)[O-])C=C(C2=O)S(=O)(=O)[O-])[As](=O)(O)O.[Na+].[Na+]
Indicator
Packaging Glass bottle
Quantity 5g
Color Undesignated
Infrared Spectrum Authentic
Suitability suitable for determination of barium
UN Number 3465
Formula Weight 576.29
Physical Form Powder
Grade Pure
Chemical Name or Material Thorin I, Pure

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

Danger