Learn More
THORLABS INC Thorlabs Pinhole Kit, SM1-Threaded, Stainless Steel Foils, Ø30 μm to Ø500 μm
Supplier:  THORLABS INC PKIT2K1
Encompass
This precision pinhole kit features stainless steel foils and SM1-threaded housings. The pinholes are made from stainless steel foils that are blackened on both sides for increased absorbance. The housings are black-anodized aluminum. Each kit is shipped in a protective plastic storage case. These kits are designed to meet IEC 62471-1 for photobiological safety of lamps and lamp systems, suitable for ophthalmological applications.
  • Precision pinholes in each kit covering four diameter ranges
  • Stainless steel foils are blackened on both sides for increased absorbance
  • 1 in. black-anodized aluminum housings engraved with item number
  • Packaged in protective plastic case for easy storage
  • Designed to meet IEC 62471-1 for photobiological safety of lamps and lamp systems, suitable for ophthalmological applications
Catalog No. NC3985423