Learn More
Aviva Systems Biology Corp THPO Antibody - middle region (ARP58734_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58734_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against THPO. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human THPO. Alternate names: MGC163194, MGDF, MKCSF, ML, MPLLG, TPOApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-62406