Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Three Stage Electric Height Adjustable Desk

Electric height adjustable rectangular desk includes a white lift frame and a walnut block laminate work surface.

 View more versions of this product

Catalog No. 06-001-054


Add to Cart
This item is not returnable. View return policy

Description

Description

  • Two motor, three stage system
  • Height range of 25 to 50 in. with memory controller
  • Available in 24 in. or 30 in. depths
Specifications

Specifications

60.9 cm
24 in.
63.5 to 127 cm
25 to 50 in.
60 in.
Steel
152.4 cm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit