Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-threo-3-Methylglutamic acid, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

63088-04-0
C6H11NO4
161.157
FHJNAFIJPFGZRI-WUJLRWPWSA-N
11412540

EAAT2 and EAAT4 blocker

A potent blocker of glutamate transport; selective for EAAT2 and EAAT4 (IC50 values are 90, 109, 1600 and 1080 μM for EAAT2, EAAT4, EAAT1 and EAAT3 respectively).
SMILES CC(CC(=O)O)C(C(=O)O)N
IUPAC Name (2S,3R)-2-amino-3-methylpentanedioic acid
Formula Weight 161.16
Quantity 50mg
Chemical Name or Material (±)-threo-3-Methylglutamic acid
Grade >98%

Recommended Storage : Store at Room Temperature