Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
threo Ifenprodil hemitartrate, Tocris Bioscience™
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Description
Potent σ receptor agonist (Ki values are 59.1 and 2 nM for σ1 and σ2 receptors respectively) and GluN2B (formally NR2B) subunit-selective NMDA receptor antagonist (IC50 values are 0.22 and 324.8 μM at GluN2B and GluN2A respectively). Displays ∽8-fold reduced affinity at α-adrenoceptors compared to Ifenprodil (Cat.No. 0545). Inhibits the hERG potassium channel (IC50 = 88 nM) and exhibits antiarrhythmic activity in vivo.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Quantity | 50 mg |
Formula Weight | 400.49 |
Percent Purity | >98% |
Chemical Name or Material | threo Ifenprodil hemitartrate |
Safety and Handling
Recommended Storage | Store at Room Temperature |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'