Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombomodulin/BDCA-3 Mouse anti-Human, Clone: QBEND/40, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $436.00
Specifications
Antigen | Thrombomodulin/BDCA-3 |
---|---|
Clone | QBEND/40 |
Dilution | Flow Cytometry Neat, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:100, Immunohistochemistry-Frozen 1:10-1:100, Electron Microscopy |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10064982
|
Novus Biologicals
NB10064982 |
0.1 mL |
Each for $436.00
|
|
NB017676
|
Novus Biologicals
NB100649820.025ML |
0.025 mL |
Each for $176.85
|
|
Description
Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy.Specifications
Thrombomodulin/BDCA-3 | |
Flow Cytometry Neat, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:100, Immunohistochemistry-Frozen 1:10-1:100, Electron Microscopy | |
Monoclonal | |
Supernatant | |
RUO | |
P07204 | |
7056 | |
Human endothelium (thrombomodulin) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
QBEND/40 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
THBD | |
IgG2a | |
Tissue culture supernatant | |
Recognizes all human endothelium tested. Preliminary evidence suggests that this recognizes Thrombomodulin, but does not inhibit Thrombomodulin dependent activation of protein C. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title