Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombomodulin/BDCA-3 Mouse anti-Human, Mouse, Alexa Fluor™ 350, Clone: 6C8.2F9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP242213AF350
Description
Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Thrombomodulin/BDCA-3 | |
6C8.2F9 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified, Conjugated | |
THBD | |
Partial recombinant human CD141 protein (between amino acids 200-400) [UniProt P07204]. | |
Protein G purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human, Mouse |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
Mouse | |
IgG1 Lambda | |
0.1 mL | |
Primary | |
7056.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title