Learn More
Thrombomodulin/BDCA-3 Mouse anti-Human, Mouse, Alexa Fluor™ 532, Clone: 6C8.2F9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP242213AF532
Catalog No. NP242213A32
Add to cart
Description
Description
Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Thrombomodulin/BDCA-3
6C8.2F9
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
Mouse
IgG1 Lambda
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
THBD
Partial recombinant human CD141 protein (between amino acids 200-400) [UniProt P07204].
0.1 mL
Primary
7056