Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombomodulin/BDCA-3 Mouse anti-Human, PE/Atto594, Clone: THBD/1591, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254490PEATT594

Catalog No. N254490P594


Add to cart

Description

Description

Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Thrombomodulin/BDCA-3
THBD/1591
Flow Cytometry
Conjugated, Purified
THBD
Recombinant human Thrombomodulin protein fragment (aa69-194) (exact sequence is proprietary)
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
Mouse
IgG2b Kappa
0.1 mL
Primary
7056.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit