Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombomodulin/BDCA-3 Rabbit anti-Rat, DyLight 405, Clone: 15, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290919V

Catalog No. NB041364


Add to cart

Description

Description

Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Thrombomodulin/BDCA-3
15
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Rat
Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
Recombinant Rat Thrombomodulin/BDCA-3 Protein
Protein A purified
RUO
Primary
7056
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit