Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombomodulin Mouse anti-Human,Rat, Unconjugated, Clone: 1009, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen Thrombomodulin
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS21561MI
View Documents
MilliporeSigma
MABS2156100UL
100 μL
Each for $386.00
Add to cart
 
MABS21562MI
View Documents
MilliporeSigma
MABS215625UL
25 μL
Each for $153.50
Add to cart
 
Description

Description

Thrombomodulin Monoclonal antibody specifically detects Thrombomodulin in Human,Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibits Activity/Function, Radioimmunoassay, Western Blotting.
Specifications

Specifications

Thrombomodulin
Unconjugated
Mouse
TM;Fetomodulin;CD141
Recombinant fragment corresponding to 241 amino acids corresponding to six EGF repeats from human Thrombomodulin.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays, Western Blot
Purified
Human, Rat
THBD, THRM
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit