Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombopoietin/Tpo Rabbit anti-Canine, Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | Thrombopoietin/Tpo |
---|---|
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15929320
|
Novus Biologicals
NBP15929320UL |
20 μL |
Each for $152.22
|
|
NBP159293
|
Novus Biologicals
NBP159293 |
100 μL |
Each for $436.00
|
|
Description
Thrombopoietin/THPO Polyclonal specifically detects Thrombopoietin/THPO in Human samples. It is validated for Western Blot, ELISA.Specifications
Thrombopoietin/Tpo | |
Polyclonal | |
Rabbit | |
Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
P40225 | |
7066 | |
IgG | |
Affinity Purified | |
35 kDa |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDFC-mpl ligand, ML, MPL ligand, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, thrombopoietin, thrombopoietin nirs variant 1, TPOMKCSF | |
THPO | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title