Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Thrombospondin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110483
Description
Immunogen sequence: TDDDYAGFLF SYQDSGRFYV VMWKQTEQTY WQATPFRAVA QPGLQLKAVT SVSGPGEHLR NALWHTGHTP DQVRLLWTDP RNVGWRDKTS YRWQLLHRPQ VGYIRVKLYE GPQLVADSGV IIDTSMRGGR LGVFCFSQEN IIWSNLQYRC NDTVPEDFEP FRRQLLQGRV.
Adhesive glycoprotein that mediates cell-to-cell and cell-to-matrix interactions. Can bind to fibrinogen, fibronectin, laminin and type V collagen.
Specifications
Thrombospondin 3 | |
Polyclonal | |
Unconjugated | |
THBS3 | |
THBS3; Thbs-3; thrombospondin 3; thrombospondin-3; Tsp3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
21827, 681309 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q05895 | |
THBS3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 787-956 of human THBS3 (NP_009043.1). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction