Learn More
Thrombospondin (TSP) Ab-4 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results Epredia™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody.
Supplier: Epredia MS421R1
Description
Thrombospondin is a protein from platelet a-granules. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.Host Species: Mouse
Clone: A6.1
Isotype: IgG1
Species Reactivity: Human, Cow, Pig, Horse, Dog, Sheep, Mouse and Rat. Others not known.
Epitope: Collagen Type V-binding domain of TSP
Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)
Positive Control: Tonsil or Placenta
Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
- Electron Microscopy
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Thrombospondin (TSP) Ab-4 | |
Monoclonal | |
Unconjugated | |
Mouse | |
1 mL | |
Angiogenesis | |
Bovine, Canine, Equine, Human, Mouse, Porcine, Rat, Sheep | |
IgG1 |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
A6.1 | |
Ready-to-use for IHC (UVLP) | |
Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets | |
RUO | |
Primary | |
Prediluted |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.