Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody

Provide accurate, reproducible results Thermo Scientific™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS421P0

 View more versions of this product

Catalog No. MS421P0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Provide accurate, reproducible results Thermo Scientific™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: A6.1

Isotype: IgG1

Species Reactivity: Human, Cow, Pig, Horse, Dog, Sheep, Mouse and Rat. Others not known.

Epitope: Collagen Type V-binding domain of TSP

Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets

Molecular Weight: ~450kDa (nonreduced), 170 to 180kDa (reduced)

Positive Control: Tonsil or Placenta

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Thrombospondin (TSP) Ab-4
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG1
RUO
Primary
Monoclonal
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
A6.1
Unconjugated
Concentrated
Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
100μL
Angiogenesis
Primary
Bovine, Canine, Equine, Human, Murine, Porcine, Rat, Sheep
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit