Learn More
THSD4 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 206191AP
Catalog No. 50-173-2989
Add to cart
Specifications
Specifications
THSD4
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZMP0
ADAMTSL-6, THSD4
Rabbit
IgG
150 μL
-20°C
Primary
79875
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
THSD4
Liquid
THSD4
THSD4 Fusion Protein Ag14682
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human