Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thujone Standard Mixture, MilliporeSigma™ Supelco™
$106.00 - $414.00
Specifications
Description | Reference substance for GC |
---|---|
CAS | 76231-76-0 |
Type | Analytical Standard |
Linear Formula | C10H16 O |
MDL Number | MFCD00135597 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-9598
|
MilliporeSigma Supelco
043141MLF |
1 mL |
Each for $106.00
|
|
|||||
11-101-8283
|
MilliporeSigma Supelco
043145MLF |
5 mL |
Each for $414.00
|
|
|||||
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal CareSpecifications
Reference substance for GC | |
Analytical Standard | |
MFCD00135597 | |
152.23 | |
β-thujone, 2-10% | |
84°C to 86°C (17 mmHg) |
76231-76-0 | |
C10H16 O | |
alpha,beta-Thujone; (-)-1-Isopropyl-4-methylbicyclo[3.1.0 ]hexan-3-one | |
Analytical Standard | |
0.925 g/mL (at 25°C) | |
67°C |
Safety and Handling
Recommended Storage : 2°C to 8°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title